Product Details

SNP ID
rs2374320
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.7:151958254 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
ACGCAGCAGGCTGCACTCGGCTGGC[A/G]CTACCGACCTAGATTCTGCACACAC
Phenotype
MIM: 615133
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
GALNTL5 PubMed Links

Gene Details

Gene
GALNTL5
Gene Name
polypeptide N-acetylgalactosaminyltransferase-like 5
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_145292.3 Intron NP_660335.2
XM_006715862.3 Intron XP_006715925.1
XM_011515849.2 Intron XP_011514151.1
XM_011515852.2 Intron XP_011514154.1
XM_011515853.2 Intron XP_011514155.1
XM_017011793.1 Intron XP_016867282.1
XM_017011794.1 Intron XP_016867283.1
XM_017011795.1 Intron XP_016867284.1
XM_017011796.1 Intron XP_016867285.1

View Full Product Details