Transcript Accession | SNP Location | SNP Type | Codon Change | Amino Acid Change | Protein ID |
---|---|---|---|---|---|
NM_001243286.1 | Intron | NP_001230215.1 | |||
NM_001243288.1 | Intron | NP_001230217.1 | |||
NM_015480.2 | Intron | NP_056295.1 | |||
XM_011512663.1 | Intron | XP_011510965.1 | |||
XM_017006123.1 | Intron | XP_016861612.1 | |||
XM_017006124.1 | Intron | XP_016861613.1 | |||
XM_017006125.1 | Intron | XP_016861614.1 | |||
XM_017006126.1 | Intron | XP_016861615.1 | |||
XM_017006127.1 | Intron | XP_016861616.1 |