Product Details

SNP ID
rs2399731
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.5:121966379 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
TTGGTACTTTGAAGAGAGAATTTTT[A/G]ATTGCTTTTATAGCAGCTTCCCTAA
Phenotype
MIM: 610479
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
SRFBP1 PubMed Links

Gene Details

Gene
SRFBP1
Gene Name
serum response factor binding protein 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_152546.2 Intron NP_689759.2
XM_017009111.1 Intron XP_016864600.1

View Full Product Details