Product Details

SNP ID
rs3087335
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.12:55967052 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GTGGAAAAGATCGGAGAGGGCACGT[A/C]CGGAGTTGTGTACAAAGCCAGAAAC
Phenotype
MIM: 116953 MIM: 155550 MIM: 179514
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
CDK2 PubMed Links

Gene Details

Gene
CDK2
Gene Name
cyclin dependent kinase 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001290230.1 284 Missense Mutation TAC,TCC Y15S NP_001277159.1
NM_001798.4 284 Missense Mutation TAC,TCC Y15S NP_001789.2
NM_052827.3 284 Missense Mutation TAC,TCC Y15S NP_439892.2
XM_011537732.1 284 Missense Mutation TAC,TCC Y15S XP_011536034.1
Gene
PMEL
Gene Name
premelanosome protein
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001200053.1 284 Intron NP_001186982.1
NM_001200054.1 284 Intron NP_001186983.1
NM_001320121.1 284 Intron NP_001307050.1
NM_001320122.1 284 Intron NP_001307051.1
NM_006928.4 284 Intron NP_008859.1
XM_006719569.1 284 Intron XP_006719632.1
XM_011538685.1 284 Intron XP_011536987.1
Gene
RAB5B
Gene Name
RAB5B, member RAS oncogene family
There are no transcripts associated with this gene.

View Full Product Details