Product Details

SNP ID
rs3087494
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.4:109710465 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
TAGAGAGAGTTGGTTTTAGGCTAAC[A/G]ATTTTTTTTCTTCTTTCCAAGACAG
Phenotype
MIM: 601532 MIM: 611652
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
CASP6 PubMed Links

Gene Details

Gene
CASP6
Gene Name
caspase 6
There are no transcripts associated with this gene.

Gene
PLA2G12A
Gene Name
phospholipase A2 group XIIA
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_030821.4 4759 UTR 3 NP_110448.2

View Full Product Details