Product Details

SNP ID
rs3087519
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.16:10929351 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
AACTCTGGCTGGGTTCCTGTAAACA[C/T]CCATCGCAGCTGCAAATAATCAGAA
Phenotype
MIM: 600005
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
CIITA PubMed Links

Gene Details

Gene
CIITA
Gene Name
class II, major histocompatibility complex, transactivator
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_000246.3 1809 Intron NP_000237.2
NM_001286402.1 1809 Intron NP_001273331.1
NM_001286403.1 1809 Intron NP_001273332.1
XM_006720880.3 1809 Intron XP_006720943.2
XM_011522484.2 1809 Intron XP_011520786.1
XM_011522485.2 1809 Intron XP_011520787.1
XM_011522486.2 1809 Intron XP_011520788.1
XM_011522487.2 1809 Intron XP_011520789.1
XM_011522488.2 1809 Intron XP_011520790.1
XM_011522489.2 1809 Intron XP_011520791.1
XM_011522490.2 1809 Intron XP_011520792.1
XM_011522491.2 1809 Intron XP_011520793.1
XM_011522492.2 1809 Intron XP_011520794.1
XM_011522493.2 1809 Intron XP_011520795.1
XM_011522494.2 1809 Intron XP_011520796.1
XM_011522495.2 1809 Intron XP_011520797.1
XM_011522496.2 1809 Intron XP_011520798.1
Gene
DEXI
Gene Name
Dexi homolog
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_014015.3 1809 Intron NP_054734.2
XM_011522455.2 1809 Intron XP_011520757.1
XM_017023171.1 1809 UTR 3 XP_016878660.1

View Full Product Details