Product Details

SNP ID
rs2447435
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.18:78984831 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AATTAAAGGGAGGCGTTCTCCAGAC[C/T]GAGACGGTGTCCACCAAGGGGATCC
Phenotype
MIM: 605079
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
SALL3 PubMed Links

Gene Details

Gene
SALL3
Gene Name
spalt like transcription factor 3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_171999.3 Intron NP_741996.2

View Full Product Details