Product Details

SNP ID
rs2430491
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.2:37203964 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
TGGACATTTAGGTTTTGTCTACTTT[C/T]TGGCATTAATAGTGCTGCTGTGAAC
Phenotype
MIM: 612828
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
CEBPZ PubMed Links

Gene Details

Gene
CEBPZ
Gene Name
CCAAT/enhancer binding protein zeta
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_005760.2 2404 Intron NP_005751.2
Gene
CEBPZOS
Gene Name
CEBPZ opposite strand
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001322373.1 2404 UTR 3 NP_001309302.1
NM_001322374.1 2404 UTR 3 NP_001309303.1
NM_001322375.1 2404 Intron NP_001309304.1
NM_001322376.1 2404 UTR 3 NP_001309305.1
XM_017003106.1 2404 UTR 3 XP_016858595.1
XM_017003107.1 2404 UTR 3 XP_016858596.1
XM_017003108.1 2404 Intron XP_016858597.1
XM_017003109.1 2404 Intron XP_016858598.1

View Full Product Details