Product Details

SNP ID
rs2167359
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:58044069 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
ACATTTATGAAATGCCTACCATATG[A/G]CAAATGTTAAACTAAGTACAGGATT
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
OR9Q1 PubMed Links

Gene Details

Gene
OR9Q1
Gene Name
olfactory receptor family 9 subfamily Q member 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001005212.3 Intron NP_001005212.1
Gene
VN2R9P
Gene Name
vomeronasal 2 receptor 9 pseudogene
There are no transcripts associated with this gene.

View Full Product Details