Product Details
- SNP ID
-
rs2421173
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.10:123136691 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- CGGTGCTTCCCGGGAAAAGTGGCCT[A/C]GGGCGTGAATAGGGAGGCTTCGGAC
- Phenotype
-
MIM: 600647
MIM: 613380
- Polymorphism
- A/C, Transversion Substitution
- Allele Nomenclature
-
- Literature Links
-
HMX2
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs115520422] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- HMX2
- Gene Name
- H6 family homeobox 2
There are no transcripts associated with this gene.
- Gene
- HMX3
- Gene Name
- H6 family homeobox 3
View Full Product Details