Product Details

SNP ID
rs2259685
Assay Type
Functionally tested
NCBI dbSNP Submissions
38
Location
Chr.1:89993292 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
CATGTTTTAGAATTCTTTTTTTTTT[C/T]TCTAGGCAAGAACTTTAATGAATTG
Phenotype
MIM: 614601
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
GEMIN8P4 PubMed Links

Gene Details

Gene
GEMIN8P4
Gene Name
gem nuclear organelle associated protein 8 pseudogene 4
There are no transcripts associated with this gene.

Gene
ZNF326
Gene Name
zinc finger protein 326
There are no transcripts associated with this gene.

View Full Product Details