Product Details

SNP ID
rs2797826
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.9:133764277 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
GTGGAACCATTGCCTGGGAGTAAGA[A/G]CAACCCAGAAAAACACCACCCTTGG
Phenotype
MIM: 600428
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
VAV2 PubMed Links

Gene Details

Gene
VAV2
Gene Name
vav guanine nucleotide exchange factor 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001134398.1 Intron NP_001127870.1
NM_003371.3 Intron NP_003362.2
XM_005272213.1 Intron XP_005272270.1
XM_017015108.1 Intron XP_016870597.1
XM_017015109.1 Intron XP_016870598.1
XM_017015110.1 Intron XP_016870599.1
XM_017015111.1 Intron XP_016870600.1
XM_017015112.1 Intron XP_016870601.1
XM_017015113.1 Intron XP_016870602.1

View Full Product Details