Product Details

SNP ID
rs2817026
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.6:35807374 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TCCTTTTTCCTGCTTCTAGCTCAGG[A/G]AGGCTTCTTGGTCCTTGTGCCAAGT
Phenotype
MIM: 609427
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
LHFPL5 PubMed Links

Gene Details

Gene
LHFPL5
Gene Name
lipoma HMGIC fusion partner-like 5
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_182548.3 Intron NP_872354.1

View Full Product Details