Product Details

SNP ID
rs775320459
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.4:9782986 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CCTTCTTCATCCTTAACTGCATGGT[C/T]CCTTTCTGCAGTGGACACCCCGAAG
Phenotype
MIM: 126453 MIM: 606142
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
DRD5 PubMed Links

Gene Details

Gene
DRD5
Gene Name
dopamine receptor D5
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_000798.4 1353 Silent Mutation GTC,GTT V319V NP_000789.1
Gene
SLC2A9
Gene Name
solute carrier family 2 member 9
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001001290.1 1353 Intron NP_001001290.1
NM_020041.2 1353 Intron NP_064425.2
XM_006713968.3 1353 Intron XP_006714031.1
XM_006713969.2 1353 Intron XP_006714032.1
XM_011513856.2 1353 Intron XP_011512158.1
XM_011513857.1 1353 Intron XP_011512159.1
XM_011513858.1 1353 Intron XP_011512160.1
XM_011513859.2 1353 Intron XP_011512161.1
XM_011513860.2 1353 Intron XP_011512162.1
XM_011513861.2 1353 Intron XP_011512163.1
XM_011513862.2 1353 Intron XP_011512164.1
XM_011513863.2 1353 Intron XP_011512165.1
XM_011513864.2 1353 Intron XP_011512166.1
XM_011513865.2 1353 Intron XP_011512167.1
XM_011513866.2 1353 Intron XP_011512168.1
XM_011513867.2 1353 Intron XP_011512169.1
XM_011513868.2 1353 Intron XP_011512170.1
XM_017008457.1 1353 Intron XP_016863946.1
XM_017008458.1 1353 Intron XP_016863947.1
XM_017008459.1 1353 Intron XP_016863948.1
XM_017008460.1 1353 Intron XP_016863949.1

View Full Product Details