Product Details

SNP ID
rs2111212
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.12:108646018 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
TCTGTGCTTTGACGCAGCCTTGGTG[A/C]GAGATGGAGAGGCAGGGCAGGTTAG
Phenotype
MIM: 605269
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
CORO1C PubMed Links

Gene Details

Gene
CORO1C
Gene Name
coronin 1C
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001105237.2 3018 UTR 3 NP_001098707.1
NM_001276471.1 3018 UTR 3 NP_001263400.1
NM_014325.3 3018 UTR 3 NP_055140.1
XM_011538125.1 3018 UTR 3 XP_011536427.1
XM_017019121.1 3018 UTR 3 XP_016874610.1
XM_017019122.1 3018 UTR 3 XP_016874611.1
XM_017019123.1 3018 UTR 3 XP_016874612.1

View Full Product Details