Product Details

SNP ID
rs2830580
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.21:26918830 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GAAAAAAAATCAGGCCAAATATACA[C/T]GACAGTTTGAATGGAATGCTGAGGA
Phenotype
MIM: 605007
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
ADAMTS5 PubMed Links

Gene Details

Gene
ADAMTS5
Gene Name
ADAM metallopeptidase with thrombospondin type 1 motif 5
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_007038.4 8745 UTR 3 NP_008969.2
XM_011529448.2 8745 UTR 3 XP_011527750.1

View Full Product Details