Product Details

SNP ID
rs2126709
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:123725219 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
TGCAACCAAGAGACTGGTTGAATAA[C/T]GGGAAGGTTAAATGCATGAGTATTT
Phenotype
MIM: 603430
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
ZNF202 PubMed Links

Gene Details

Gene
ZNF202
Gene Name
zinc finger protein 202
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001301779.1 3325 UTR 3 NP_001288708.1
NM_001301780.1 3325 UTR 3 NP_001288709.1
NM_001301819.1 3325 UTR 3 NP_001288748.1
NM_003455.3 3325 UTR 3 NP_003446.2
XM_005271659.1 3325 UTR 3 XP_005271716.1
XM_005271660.1 3325 UTR 3 XP_005271717.1
XM_005271661.1 3325 UTR 3 XP_005271718.1
XM_005271664.1 3325 UTR 3 XP_005271721.1
XM_006718901.2 3325 UTR 3 XP_006718964.1
XM_011542972.1 3325 UTR 3 XP_011541274.1
XM_011542973.1 3325 UTR 3 XP_011541275.1
XM_011542975.1 3325 UTR 3 XP_011541277.1
XM_011542976.1 3325 UTR 3 XP_011541278.1
XM_017018268.1 3325 UTR 3 XP_016873757.1

View Full Product Details