Product Details

SNP ID
rs2860055
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.4:52052572 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
AGGGATTTCCCCTCTTACATAATAA[A/G]TGGAGCGGCCAGGCAAAAGAACTGT
Phenotype
MIM: 612814
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
SPATA18 PubMed Links

Gene Details

Gene
SPATA18
Gene Name
spermatogenesis associated 18
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001297608.1 Intron NP_001284537.1
NM_145263.3 Intron NP_660306.1
XM_005265716.1 Intron XP_005265773.1
XM_005265717.2 Intron XP_005265774.1
XM_005265718.1 Intron XP_005265775.1
XM_011534369.1 Intron XP_011532671.1
XM_011534370.1 Intron XP_011532672.1
XM_011534372.1 Intron XP_011532674.1
XM_017007731.1 Intron XP_016863220.1

View Full Product Details