Product Details

SNP ID
rs2168823
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.14:52561190 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
GGACTAAAATTTAAAAGACTAACCA[C/T]ACCAACTATTGGCCAAAATGTGGAT
Phenotype
MIM: 616179
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
GPR137C PubMed Links

Gene Details

Gene
GPR137C
Gene Name
G protein-coupled receptor 137C
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001099652.1 Intron NP_001093122.1
Gene
TXNDC16
Gene Name
thioredoxin domain containing 16
There are no transcripts associated with this gene.

View Full Product Details