Product Details
- SNP ID
-
rs2914571
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.19:7069870 on Build GRCh38
- Set Membership
-
HapMap
- Context Sequence [VIC/FAM]
- GAGGCCTCGGGCTGTGCGCTCAGTT[A/C]AGCCCGAAGCTGCTGTTTCCTCCGG
- Phenotype
-
- Polymorphism
- A/C, Transversion substitution
- Allele Nomenclature
-
- Literature Links
-
ZNF557
PubMed Links
Gene Details
- Gene
- ZNF557
- Gene Name
- zinc finger protein 557
View Full Product Details