Product Details

SNP ID
rs2242995
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.5:76819840 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GCTGGAAGCCTTTCTTCGGGTTCTG[C/G]CCCCTGCGTTCGAGACCCACCCGGG
Phenotype
MIM: 600933
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
F2RL1 PubMed Links

Gene Details

Gene
F2RL1
Gene Name
F2R like trypsin receptor 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
XM_017009223.1 Intron XP_016864712.1

View Full Product Details