Product Details

SNP ID
rs2193596
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.14:74767398 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
ACAGTCTGTTCCCAAACAATGTTTT[G/T]TTGGTGTGTTCAAGTCAGAGTCCAA
Phenotype
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
YLPM1 PubMed Links

Gene Details

Gene
YLPM1
Gene Name
YLP motif containing 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_019589.2 Intron NP_062535.2
XM_005267860.3 Intron XP_005267917.1
XM_011536966.2 Intron XP_011535268.1
XM_011536967.1 Intron XP_011535269.1

View Full Product Details