Product Details

SNP ID
rs2193621
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.2:171436215 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
ATCAATTACCACATTTTACGTATCC[G/A]TTTATCAGTTGTTAGACATTGGTGT
Phenotype
MIM: 612515 MIM: 609525
Polymorphism
G/A, Transition substitution
Allele Nomenclature
Literature Links
DCAF17 PubMed Links

Gene Details

Gene
DCAF17
Gene Name
DDB1 and CUL4 associated factor 17
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001164821.1 Intron NP_001158293.1
NM_025000.3 Intron NP_079276.2
XM_006712767.1 Intron XP_006712830.1
XM_006712768.1 Intron XP_006712831.1
XM_011511881.1 Intron XP_011510183.1
XM_011511882.1 Intron XP_011510184.1
XM_017004995.1 Intron XP_016860484.1
XM_017004996.1 Intron XP_016860485.1
XM_017004997.1 Intron XP_016860486.1
XM_017004998.1 Intron XP_016860487.1
XM_017004999.1 Intron XP_016860488.1
XM_017005000.1 Intron XP_016860489.1
XM_017005001.1 Intron XP_016860490.1
XM_017005002.1 Intron XP_016860491.1
Gene
METTL8
Gene Name
methyltransferase like 8
There are no transcripts associated with this gene.

View Full Product Details