Product Details

SNP ID
rs2916116
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.12:49100132 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TCACCCAGGAGAAAGATTTAGAAAA[A/G]TGTGCTCCCTGACCAAAAAGTAGAA
Phenotype
MIM: 605423 MIM: 610007
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
DHH PubMed Links

Gene Details

Gene
DHH
Gene Name
desert hedgehog
There are no transcripts associated with this gene.

Gene
LMBR1L
Gene Name
limb development membrane protein 1 like
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001300750.1 Intron NP_001287679.1
NM_001300751.1 Intron NP_001287680.1
NM_018113.3 Intron NP_060583.2
XM_011538563.2 Intron XP_011536865.1
XM_011538564.2 Intron XP_011536866.1
XM_011538565.2 Intron XP_011536867.1
XM_011538566.2 Intron XP_011536868.1
XM_017019626.1 Intron XP_016875115.1
XM_017019627.1 Intron XP_016875116.1
XM_017019628.1 Intron XP_016875117.1
XM_017019629.1 Intron XP_016875118.1
XM_017019630.1 Intron XP_016875119.1
XM_017019631.1 Intron XP_016875120.1

View Full Product Details