Product Details

SNP ID
rs2209586
Assay Type
Functionally Tested
NCBI dbSNP Submissions
NA
Location
Chr.X:46469002 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
GCTGCTGAACCAGTAAATATATAAT[A/G]TAAACATTCTGGAAACCAACAAAAT
Phenotype
MIM: 300585
Polymorphism
A/G, Transition Substitution
Allele Nomenclature
Literature Links
KRBOX4 PubMed Links

Gene Details

Gene
KRBOX4
Gene Name
KRAB box domain containing 4
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001129898.1 Intron NP_001123370.1
NM_001129899.1 Intron NP_001123371.1
NM_001129900.1 Intron NP_001123372.1
NM_017776.2 Intron NP_060246.2

View Full Product Details