Product Details

SNP ID
rs2230292
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.12:64716750 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
AGGCTGTGTGAGGTCGCTACAGAAG[A/G]TGTTTGGAAAATCTTCGAGTCCTGA
Phenotype
MIM: 607664
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
GNS PubMed Links

Gene Details

Gene
GNS
Gene Name
glucosamine (N-acetyl)-6-sulfatase
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_002076.3 1820 Silent Mutation CAC,CAT H550H NP_002067.1

View Full Product Details