Product Details

SNP ID
rs2231178
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.7:100643425 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GAATCCTCCAACAGGCCCCAACCAC[C/T]CCTTGGGTTCACATCCCTCTGAAGC
Phenotype
MIM: 612458 MIM: 604720
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
ACTL6B PubMed Links

Gene Details

Gene
ACTL6B
Gene Name
actin like 6B
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_016188.4 Intron NP_057272.1
Gene
TFR2
Gene Name
transferrin receptor 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001206855.1 Intron NP_001193784.1
NM_003227.3 Intron NP_003218.2
XM_005250553.4 Intron XP_005250610.1
XM_017012573.1 Intron XP_016868062.1

View Full Product Details