Product Details

SNP ID
rs2231405
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.9:132884256 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
ATGAGTCCTTCATTCGTTTCTCTAC[C/T]AAATATGTAAGTGAATAACAACAAG
Phenotype
MIM: 615365 MIM: 605284
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
AK8 PubMed Links

Gene Details

Gene
AK8
Gene Name
adenylate kinase 8
There are no transcripts associated with this gene.

Gene
SPACA9
Gene Name
sperm acrosome associated 9
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001316897.1 Intron NP_001303826.1
NM_001316898.1 Intron NP_001303827.1
NM_001316900.1 Intron NP_001303829.1
NM_018956.4 Intron NP_061829.3
XM_017014231.1 Intron XP_016869720.1
Gene
TSC1
Gene Name
tuberous sclerosis 1
There are no transcripts associated with this gene.

View Full Product Details