Product Details

SNP ID
rs2231992
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:18919762 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
CAAGGATGGCAGGCTTCGCGGAGCT[C/T]GGGCTGTCATCGTGGCTCGTGGAAC
Phenotype
MIM: 606942 MIM: 604800
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
COPE PubMed Links

Gene Details

Gene
COPE
Gene Name
coatomer protein complex subunit epsilon
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_007263.3 88 Intron NP_009194.2
NM_199442.1 88 Intron NP_955474.1
NM_199444.1 88 Intron NP_955476.1
XM_011527655.1 88 Intron XP_011525957.1
XM_011527656.1 88 Intron XP_011525958.1
Gene
DDX49
Gene Name
DEAD-box helicase 49
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_019070.4 88 Silent Mutation CTC,CTT L7L NP_061943.2
XM_011528083.1 88 UTR 5 XP_011526385.1
XM_011528084.2 88 UTR 5 XP_011526386.1
Gene
HOMER3
Gene Name
homer scaffolding protein 3
There are no transcripts associated with this gene.

View Full Product Details