Product Details

SNP ID
rs2232053
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.14:69399304 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
AACCACCACACCTGTTTAAAGAACC[C/T]AAGCACCATTTAAAGCCACTGGAAA
Phenotype
MIM: 601191
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
ERH PubMed Links

Gene Details

Gene
ERH
Gene Name
enhancer of rudimentary homolog (Drosophila)
There are no transcripts associated with this gene.

Gene
SLC39A9
Gene Name
solute carrier family 39 member 9
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001252148.1 637 UTR 5 NP_001239077.1
NM_001252150.1 637 UTR 5 NP_001239079.1
NM_001252151.1 637 Intron NP_001239080.1
NM_001252152.1 637 Intron NP_001239081.1
NM_018375.4 637 UTR 5 NP_060845.2
XM_005267834.1 637 UTR 5 XP_005267891.1
XM_017021426.1 637 UTR 5 XP_016876915.1

View Full Product Details