Product Details
- SNP ID
-
rs2232083
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.20:6077323 on Build GRCh38
- Set Membership
-
HapMap
- Context Sequence [VIC/FAM]
- TCAGGCAGGTGAAAGCAGTAAAGAC[A/G]TTTTGGTCAAACTCGATGACCACCT
- Phenotype
-
MIM: 607900
- Polymorphism
- A/G, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
FERMT1
PubMed Links
Gene Details
- Gene
- FERMT1
- Gene Name
- fermitin family member 1
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_017671.4 |
2673 |
Silent Mutation |
AAC,AAT |
N628N |
NP_060141.3 |
View Full Product Details