Product Details
- SNP ID
-
rs2232122
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.7:33727738 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- ACACAAACATGCTCCTGCTCCTGGC[A/G]GAGGCAGAGCTGCTGGGAAAGACAT
- Phenotype
-
- Polymorphism
- A/G, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
FLJ20712
PubMed Links
Gene Details
- Gene
- FLJ20712
- Gene Name
- uncharacterized FLJ20712
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
XM_017012879.1 |
1292 |
Missense Mutation |
AGA,GGA |
R37G |
XP_016868368.1 |
View Full Product Details