Product Details

SNP ID
rs2232157
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.7:91264657 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GGCGGGACACGACCCCGGCGCGCCC[G/T]AGCCACCCGGGTTCTCCCCGCCGCC
Phenotype
MIM: 603408
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
FZD1 PubMed Links

Gene Details

Gene
FZD1
Gene Name
frizzled class receptor 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_003505.1 190 UTR 5 NP_003496.1

View Full Product Details