Product Details

SNP ID
rs2232224
Assay Type
Functionally tested
NCBI dbSNP Submissions
52
Location
Chr.1:42163369 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
TCAAGTGCCACCACAGTATTCCCGA[A/G]CCCCACCAATCAGAGAGGAAGGAGG
Phenotype
MIM: 139392 MIM: 601271
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
GUCA2A PubMed Links

Gene Details

Gene
GUCA2A
Gene Name
guanylate cyclase activator 2A
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_033553.2 Intron NP_291031.2
Gene
GUCA2B
Gene Name
guanylate cyclase activator 2B
There are no transcripts associated with this gene.

View Full Product Details