Product Details
- SNP ID
-
rs2232254
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.20:9514281 on Build GRCh38
- Set Membership
-
HapMap
- Context Sequence [VIC/FAM]
- TCAGGAAACTGTTGCCATAGCATTT[C/T]CCCTTCTCTATTCCTGCATAGCCCT
- Phenotype
-
MIM: 614641
- Polymorphism
- C/T, Transition Substitution
- Allele Nomenclature
-
- Literature Links
-
LAMP5
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs150359838,rs543756911] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- LAMP5
- Gene Name
- lysosomal associated membrane protein family member 5
- Gene
- LAMP5-AS1
- Gene Name
- LAMP5 antisense RNA 1
There are no transcripts associated with this gene.
View Full Product Details