Product Details

SNP ID
rs2067850
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.17:69249743 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
CCATAGTTTCTAAAATAAACCGAGT[A/C]CCCAGTTTTAAACTTTTTAAATAAA
Phenotype
MIM: 612508 MIM: 612503
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
ABCA10 PubMed Links

Gene Details

Gene
ABCA10
Gene Name
ATP binding cassette subfamily A member 10
There are no transcripts associated with this gene.

Gene
ABCA5
Gene Name
ATP binding cassette subfamily A member 5
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_018672.4 Intron NP_061142.2
NM_172232.3 Intron NP_758424.1

View Full Product Details