Product Details

SNP ID
rs2070352
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.21:34185855 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
TGAGGAGTTACCGGTTGAATTTTGT[C/G]CCCCTCAACAAAGATGTTGAAGTCC
Phenotype
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
LINC00310 PubMed Links

Gene Details

Gene
LINC00310
Gene Name
long intergenic non-protein coding RNA 310
There are no transcripts associated with this gene.

View Full Product Details