Product Details

SNP ID
rs2807663
Assay Type
Functionally tested
NCBI dbSNP Submissions
80
Location
Chr.1:214613292 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
CCCAGGTCTTCATCATCTTCTAAAC[A/C]CCAGCTAACTATGCCACCTCCTACC
Phenotype
MIM: 600236
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
CENPF PubMed Links

Gene Details

Gene
CENPF
Gene Name
centromere protein F
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_016343.3 Intron NP_057427.3
XM_011509082.2 Intron XP_011507384.1
XM_017000086.1 Intron XP_016855575.1

View Full Product Details