Product Details

SNP ID
rs2808417
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.9:123358030 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
GCCCAGGGGCAGCCTTTCCTCTGCC[A/G]TTTCTGTGCTGGGAGCGCCATCCCT
Phenotype
MIM: 609720
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
CRB2 PubMed Links

Gene Details

Gene
CRB2
Gene Name
crumbs 2, cell polarity complex component
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_173689.6 Intron NP_775960.4
XM_005251934.2 Intron XP_005251991.1
XM_011518556.2 Intron XP_011516858.1
XM_011518557.2 Intron XP_011516859.1
XM_011518558.2 Intron XP_011516860.1

View Full Product Details