Product Details

SNP ID
rs2076593
Assay Type
Functionally tested
NCBI dbSNP Submissions
37
Location
Chr.1:111725366 on Build GRCh38
Set Membership
HapMap JSNP
Context Sequence [VIC/FAM]
ATCCACATTGTCTTTTCCCAGCCAG[C/T]GAGTGTTCCCTACTGGAGTGGGAGG
Phenotype
MIM: 179520
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
FAM212B PubMed Links

Gene Details

Gene
FAM212B
Gene Name
family with sequence similarity 212 member B
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_019099.4 2774 UTR 3 NP_061972.1
NM_198926.2 2774 UTR 3 NP_945120.1
XM_011541783.2 2774 UTR 3 XP_011540085.1
Gene
RAP1A
Gene Name
RAP1A, member of RAS oncogene family
There are no transcripts associated with this gene.

View Full Product Details