Product Details

SNP ID
rs2071036
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:62852565 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
ATTGGCTCCCAGTGTCTTAATCTGT[A/G]TCTGCTAACACTTTAAGGTACATCT
Phenotype
MIM: 158070 MIM: 603222 MIM: 603223 MIM: 603224 MIM: 603225 MIM: 603226 MIM: 603227 MIM: 603228 MIM: 603229 MIM: 603230
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
SLC3A2 PubMed Links

Gene Details

Gene
SLC3A2
Gene Name
solute carrier family 3 member 2
There are no transcripts associated with this gene.

Gene
SNHG1
Gene Name
small nucleolar RNA host gene 1
There are no transcripts associated with this gene.

Gene
SNORD22
Gene Name
small nucleolar RNA, C/D box 22
There are no transcripts associated with this gene.

Gene
SNORD25
Gene Name
small nucleolar RNA, C/D box 25
There are no transcripts associated with this gene.

Gene
SNORD26
Gene Name
small nucleolar RNA, C/D box 26
There are no transcripts associated with this gene.

Gene
SNORD27
Gene Name
small nucleolar RNA, C/D box 27
There are no transcripts associated with this gene.

Gene
SNORD28
Gene Name
small nucleolar RNA, C/D box 28
There are no transcripts associated with this gene.

Gene
SNORD29
Gene Name
small nucleolar RNA, C/D box 29
There are no transcripts associated with this gene.

Gene
SNORD30
Gene Name
small nucleolar RNA, C/D box 30
There are no transcripts associated with this gene.

Gene
SNORD31
Gene Name
small nucleolar RNA, C/D box 31
There are no transcripts associated with this gene.

View Full Product Details