Product Details

SNP ID
rs2071429
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.X:154532293 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
ACGGAGCTCGTCGCTGAGGGGACAT[A/G]GTATGGCTTGGGAGGCCGGTGGCAC
Phenotype
MIM: 305900 MIM: 300248
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
G6PD PubMed Links

Gene Details

Gene
G6PD
Gene Name
glucose-6-phosphate dehydrogenase
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_000402.4 Intron NP_000393.4
NM_001042351.2 Intron NP_001035810.1
Gene
IKBKG
Gene Name
inhibitor of kappa light polypeptide gene enhancer in B-cells, kinase gamma
There are no transcripts associated with this gene.

View Full Product Details