Product Details

SNP ID
rs2071778
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.X:47483855 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
GCAGAAAATGTTACAGGTCTTAACA[C/T]CATTTTGCATTTCCGCAAACGAACA
Phenotype
MIM: 314995
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
LINC01560 PubMed Links

Gene Details

Gene
LINC01560
Gene Name
long intergenic non-protein coding RNA 1560
There are no transcripts associated with this gene.

Gene
ZNF41
Gene Name
zinc finger protein 41
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001324139.1 Intron NP_001311068.1
NM_001324140.1 Intron NP_001311069.1
NM_001324141.1 Intron NP_001311070.1
NM_001324142.1 Intron NP_001311071.1
NM_001324143.1 Intron NP_001311072.1
NM_001324144.1 Intron NP_001311073.1
NM_001324145.1 Intron NP_001311074.1
NM_001324147.1 Intron NP_001311076.1
NM_001324148.1 Intron NP_001311077.1
NM_001324149.1 Intron NP_001311078.1
NM_001324150.1 Intron NP_001311079.1
NM_001324151.1 Intron NP_001311080.1
NM_001324152.1 Intron NP_001311081.1
NM_001324153.1 Intron NP_001311082.1
NM_001324154.1 Intron NP_001311083.1
NM_001324155.1 Intron NP_001311084.1
NM_001324156.1 Intron NP_001311085.1
NM_001324157.1 Intron NP_001311086.1
NM_007130.3 Intron NP_009061.1
NM_153380.3 Intron NP_700359.1
XM_006724550.3 Intron XP_006724613.1
XM_006724555.3 Intron XP_006724618.1
XM_017029810.1 Intron XP_016885299.1
XM_017029811.1 Intron XP_016885300.1
XM_017029812.1 Intron XP_016885301.1
XM_017029813.1 Intron XP_016885302.1
XM_017029814.1 Intron XP_016885303.1
XM_017029815.1 Intron XP_016885304.1
XM_017029816.1 Intron XP_016885305.1
XM_017029817.1 Intron XP_016885306.1

View Full Product Details