Product Details

SNP ID
rs2072103
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:1105539 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GGCTGCTGCGTCCCCGGGGCCCGCA[A/G]AGGCGGGTGGGTGGGGGTCGGGGTG
Phenotype
MIM: 138322 MIM: 615729
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
GPX4 PubMed Links

Gene Details

Gene
GPX4
Gene Name
glutathione peroxidase 4
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001039847.2 Intron NP_001034936.1
NM_001039848.2 Intron NP_001034937.1
NM_002085.4 Intron NP_002076.2
Gene
SBNO2
Gene Name
strawberry notch homolog 2
There are no transcripts associated with this gene.

View Full Product Details