Product Details

SNP ID
rs2072269
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:15913929 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
CAGTTTCTGGGGGCAAAAGTGAGGG[A/G]ATCTGACTGTGCCCATGACCCCCAT
Phenotype
MIM: 611517
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
CYP4F11 PubMed Links

Gene Details

Gene
CYP4F11
Gene Name
cytochrome P450 family 4 subfamily F member 11
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001128932.1 Intron NP_001122404.1
NM_021187.3 Intron NP_067010.3

View Full Product Details