Product Details
- SNP ID
-
rs2240744
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.19:5455724 on Build GRCh38
- Set Membership
-
HapMap
- Context Sequence [VIC/FAM]
- ACCATGGGGTGGCCACGGCCGGGCC[A/G]AGCCCTCGTGGCAGTCAAAGCCTTG
- Phenotype
-
MIM: 612063
- Polymorphism
- A/G, Transition Substitution
- Allele Nomenclature
-
- Literature Links
-
ZNRF4
PubMed Links
Gene Details
- Gene
- ZNRF4
- Gene Name
- zinc and ring finger 4
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_181710.3 |
310 |
Missense Mutation |
CAA,CGA |
Q78R |
NP_859061.3 |
View Full Product Details