Product Details

SNP ID
rs2240821
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.7:139779626 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
GGTCATTTATCTCCTTCCCTTCTAT[C/T]CTCCCTGCAATCTTTAACTTTAGTC
Phenotype
MIM: 606868 MIM: 274180
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
HIPK2 PubMed Links

Gene Details

Gene
HIPK2
Gene Name
homeodomain interacting protein kinase 2
There are no transcripts associated with this gene.

Gene
TBXAS1
Gene Name
thromboxane A synthase 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001061.4 Intron NP_001052.2
NM_001130966.2 Intron NP_001124438.1
NM_001166253.1 Intron NP_001159725.1
NM_001166254.1 Intron NP_001159726.1
NM_001314028.1 Intron NP_001300957.1
NM_030984.3 Intron NP_112246.2
XM_011516544.2 Intron XP_011514846.1
XM_017012569.1 Intron XP_016868058.1
XM_017012570.1 Intron XP_016868059.1
XM_017012571.1 Intron XP_016868060.1
XM_017012572.1 Intron XP_016868061.1

View Full Product Details