Product Details
- SNP ID
-
rs2267434
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.22:41482591 on Build GRCh38
- Set Membership
-
HapMap
- Context Sequence [VIC/FAM]
- TTTTGTGGCTGAGGAAGTGCTATAA[C/T]GATGCTTAGGATGGAGAAGACATAA
- Phenotype
-
MIM: 100850
- Polymorphism
- C/T, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
ACO2
PubMed Links
Gene Details
- Gene
- ACO2
- Gene Name
- aconitase 2
View Full Product Details