Product Details

SNP ID
rs2241181
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.12:12118789 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CTGCCTTCAAATAATTTTAAACAAA[C/T]TGGCACACAAATTAGTATTTTGAAG
Phenotype
MIM: 603507
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
LOC100506248 PubMed Links

Gene Details

Gene
LOC100506248
Gene Name
prothymosin alpha
There are no transcripts associated with this gene.

Gene
LRP6
Gene Name
LDL receptor related protein 6
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_002336.2 7321 UTR 3 NP_002327.2
XM_006719078.3 7321 UTR 3 XP_006719141.1
XM_011520671.2 7321 UTR 3 XP_011518973.1
XM_017019304.1 7321 UTR 3 XP_016874793.1
Gene
MIR1244-4
Gene Name
microRNA 1244-4
There are no transcripts associated with this gene.

View Full Product Details