Product Details

SNP ID
rs2241300
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.8:30995952 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GTGTGCAGTTAGCATCCACAAAAGC[A/G]CCATTGGGTATGGGAGAATTAGCAC
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
PURG PubMed Links

Gene Details

Gene
PURG
Gene Name
purine rich element binding protein G
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001015508.2 2543 UTR 3 NP_001015508.1
NM_001323311.1 2543 Intron NP_001310240.1
NM_001323312.1 2543 UTR 3 NP_001310241.1
NM_013357.2 2543 Intron NP_037489.1

View Full Product Details